Phosphoenolpyruvic acid tris(cyclohexylammonium)salt, hydrate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

P0256-1G P0256-5G
TCIAP0256-1GEA 103 EUR
TCIAP0256-1G TCIAP0256-5G
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt, hydrate ≥98.0% (by titrimetric analysis)
Phosphoenolpyruvic acid tris(cyclohexylammonium)salt
Kaava: C₃H₅O₆P·3C₆H₁₄N·xH₂O
Storage Temperature: Jääkaappi
MDL numero: MFCD00004258
CAS-numero: 35556-70-8
EINECS: 252-618-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Methanol almost transparency
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR