Phenoxyacetyl chloride ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Vaara

P0113-25G P0113-500G
TCIAP0113-25GEA 37.4 EUR
TCIAP0113-25G TCIAP0113-500G
Phenoxyacetyl chloride ≥98.0% (by GC, titration analysis)
Phenoxyacetyl chloride
Kaava: C₈H₇ClO₂
Kiehumispiste: 226°C
Sulamispiste: 28…30 °C
Tiheys: 1,23 g/cm³ (20 °C)
Leimahduspiste: 108°C
MDL numero: MFCD00000726
CAS-numero: 701-99-5
EINECS: 211-862-4
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e51de2b21f7d547d1cc93782c33d7e580043a603&vid=e51de2b21f7d547d1cc93782c33d7e580043a603&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR