1,3,5-Pentanetricarboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

P0075-25G P0075-5G
TCIAP0075-25GEA 456 EUR
TCIAP0075-25G TCIAP0075-5G
1,3,5-Pentanetricarboxylic acid ≥98.0% (by GC, titration analysis)
1,3,5-Pentanetricarboxylic acid
Kaava: C₈H₁₂O₆
MDL numero: MFCD00020546
CAS-numero: 6940-58-5
EINECS: 230-087-2

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR