3-Oxo-N-(4-trifluoromethylphenyl)butyramide ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI

O0444-200MG O0444-1G
TCIAO0444-200MGEA 90.2 EUR
TCIAO0444-200MG TCIAO0444-1G
3-Oxo-N-(4-trifluoromethylphenyl)butyramide ≥98.0% (by HPLC, total nitrogen)
3-Oxo-N-(4-trifluoromethylphenyl)butyramide
Kaava: C₁₁H₁₀F₃NO₂
MDL numero: MFCD00231301
CAS-numero: 351-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79d4b6dc11734346fce3bdfff23cc14007ad36b7&vid=79d4b6dc11734346fce3bdfff23cc14007ad36b7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 152.0 to 156.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR