L-Ornithine L-Aspartate ≥96.0% (by titrimetric analysis)

Toimittaja: TCI

O0440-25G
TCIAO0440-25GEA 63.8 EUR
TCIAO0440-25G
L-Ornithine L-Aspartate ≥96.0% (by titrimetric analysis)
L-Ornithine L-Aspartate
Kaava: C₉H₁₉N₃O₆
Storage Temperature: Jääkaappi
MDL numero: MFCD00058084
CAS-numero: 3230-94-2
EINECS: 221-772-7
Merck Index: 14,06874
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2834d1f42d70845ff343f8599a6a12bf68aaae8&vid=d2834d1f42d70845ff343f8599a6a12bf68aaae8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 96.0 %
Specific rotation [a]20/D +26.0 to +29.0 deg (C=1, 6mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR