Ondansetron hydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Vaara

O0407-1G O0407-5G
TCIAO0407-1GEA 56.1 EUR
TCIAO0407-1G TCIAO0407-5G
Ondansetron hydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Ondansetron hydrochloride dihydrate
Kaava: C₁₈H₁₉N₃O·HCl·₂H₂O
Storage Temperature: Jääkaappi
MDL numero: MFCD00374371
CAS-numero: 103639-04-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=303b043daeeffda65d011456d5c62998ad5c7913&vid=303b043daeeffda65d011456d5c62998ad5c7913&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Melting point 176.0 to 180.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR