L(+)-Ornithine dihydrochloride ≥98.0% (by total nitrogen basis)

Toimittaja: TCI

O0089-1G O0089-5G
TCIAO0089-1GEA 17.6 EUR
TCIAO0089-1G TCIAO0089-5G
L(+)-Ornithine dihydrochloride ≥98.0% (by total nitrogen basis)
L(+)-Ornithine dihydrochloride
Kaava: C₅H₁₄Cl₂N₂O₂
MDL numero: MFCD00066571
CAS-numero: 6211-16-1
EINECS: 228-277-5
Merck Index: 14,06874

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +16.0 to +19.0 deg (C=5, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR