DL-Ornithine hydrochloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

O0063-1G
TCIAO0063-1GEA 17.6 EUR
TCIAO0063-1G
DL-Ornithine hydrochloride ≥98.0% (by titrimetric analysis)
DL-Ornithine hydrochloride
Kaava: C₅H₁₂N₂O₂·HCl
MDL numero: MFCD00065398
CAS-numero: 1069-31-4
EINECS: 213-956-0
Merck Index: 14,06874
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fff25d4a202e61b0a4d95b41b9e4a2c02fa2311c&vid=fff25d4a202e61b0a4d95b41b9e4a2c02fa2311c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR