Oleyl amine >50.0% (by GC)

Toimittaja: TCI
Vaara

O0059-25ML O0059-500ML O0059-100ML
TCIAO0059-25MLEA 17.6 EUR
TCIAO0059-25ML TCIAO0059-500ML TCIAO0059-100ML
Oleyl amine >50.0% (by GC)
Oleyl amine
Kaava: C₁₈H₃₇N
Kiehumispiste: 364 °C (1013 hPa)
Sulamispiste: 21 °C
Tiheys: 0,813 g/cm³ (20 °C)
Leimahduspiste: 158°C
MDL numero: MFCD00066507
CAS-numero: 112-90-3
EINECS: 204-015-5
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aea92ea7e4ab5fcbff6381e6d8c20b118d771162&vid=aea92ea7e4ab5fcbff6381e6d8c20b118d771162&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 50.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR