N749 Black Dye ≥85.0% (by HPLC)
Toimittaja: TCI
Varoitus
N1104-100MG
TCIAN1104-100MGEA 310 EUR
TCIAN1104-100MG
N749 Black Dye ≥85.0% (by HPLC)
N749 Black Dye
Kaava: C₆₉H₁₁₇N₉O₆RuS₃
MDL numero: MFCD02097764
CAS-numero: 359415-47-7
Order Now
Testitulosten erittely
Appearance Dark green to Dark blue to Black powder to crystal
Purity (HPLC) min. 85.0 area%
Elemental analysis (Nitrogen) 8.50 to 10.00 %
Molar extinction coefficient min. 20000 (EtOH, 325 to 335nm)