9-(1-Naphthyl)carbazole ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

N1012-1G N1012-5G
TCIAN1012-1GEA 112 EUR
TCIAN1012-1G TCIAN1012-5G
9-(1-Naphthyl)carbazole ≥98.0% (by GC)
9-(1-Naphthyl)carbazole
Kaava: C₂₂H₁₅N
MDL numero: MFCD26127424
CAS-numero: 22034-43-1

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 126.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR