4-Nitrophenyl trifluoromethanesulphonate ≥99.0% (by GC)

Toimittaja: TCI
Vaara

N0993-5G N0993-1G
TCIAN0993-5GEA 63.8 EUR
TCIAN0993-5G TCIAN0993-1G
4-Nitrophenyl trifluoromethanesulphonate ≥99.0% (by GC)
4-Nitrophenyl trifluoromethanesulphonate
Kaava: C₇H₄F₃NO₅S
MDL numero: MFCD00192354
CAS-numero: 17763-80-3
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 52.0 to 56.0 deg-C

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR