4'-Nitrobiphenyl-4-carboxaldehyde ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

N0967-1G N0967-5G
TCIAN0967-1GEA 106 EUR
TCIAN0967-1G TCIAN0967-5G
4'-Nitrobiphenyl-4-carboxaldehyde ≥98.0% (by GC)
4'-Nitrobiphenyl-4-carboxaldehyde
Kaava: C₁₃H₉NO₃
MDL numero: MFCD06200931
CAS-numero: 98648-23-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2988faebb4d8be62ee6dd92fb0d900ab5e626d4e&vid=2988faebb4d8be62ee6dd92fb0d900ab5e626d4e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR