2-Nitrophenyl-β-D-xylopyranoside ≥98.0% (by HPLC)

Toimittaja: TCI

N0868-100MG
TCIAN0868-100MGEA 57.2 EUR
TCIAN0868-100MG
2-Nitrophenyl-β-D-xylopyranoside ≥98.0% (by HPLC)
2-Nitrophenyl-beta-D-xylopyranoside
Kaava: C₁₁H₁₃NO₇
Storage Temperature: Pakastin
MDL numero: MFCD00047515
CAS-numero: 10238-27-4
EINECS: 233-571-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7ccc299c425821605c667d7973194b2e8e84dcb&vid=f7ccc299c425821605c667d7973194b2e8e84dcb&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 171.0 to 175.0 deg-C
Specific rotation [a]20/D -86.0 to -91.0 deg (C=0.6, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR