Nortropinone hydrochloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

N0844-25G N0844-5G
TCIAN0844-25GEA 307 EUR
TCIAN0844-25G TCIAN0844-5G
Nortropinone hydrochloride ≥98.0% (by titrimetric analysis)
Nortropinone hydrochloride
Kaava: C₇H₁₂ClNO
Sulamispiste: 204°C
MDL numero: MFCD03613582
CAS-numero: 25602-68-0

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR