(R)-(+)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

N0724-1G N0724-5G
TCIAN0724-1GEA 49.5 EUR
TCIAN0724-1G TCIAN0724-5G
(R)-(+)-1-(2-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)
(R)-(+)-1-(2-Naphthyl)ethylamine
Kaava: C₁₂H₁₃N
Kiehumispiste: 166°C
Sulamispiste: 53°C
MDL numero: MFCD00085367
CAS-numero: 3906-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=456e9befccf324c163261a1f21787466bd2e3f00&vid=456e9befccf324c163261a1f21787466bd2e3f00&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +20.0 to +23.0deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR