3-(2-Naphthyl)-L-alanine hydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI

Synonyms: H-2-Nal-OH·HCl, (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride

N0683-1G N0683-5G
TCIAN0683-1GEA 135 EUR
TCIAN0683-1G TCIAN0683-5G
3-(2-Naphthyl)-L-alanine hydrochloride ≥98.0% (by HPLC, titration analysis)
3-(2-Naphthyl)-L-alanine hydrochloride
Kaava: C₁₃H₁₃NO₂·HCl
MW: 251.71 g/mol
MDL numero: MFCD03095595
CAS-numero: 122745-12-4

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -8.0 to -11.0 deg (C=1, 1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR