3-Nitro-o-kresoli ≥98.0% (by GC)

Toimittaja: TCI
Vaara

N0627-25G N0627-5G
TCIAN0627-25GEA 104 EUR
TCIAN0627-25G TCIAN0627-5G
3-Nitro-o-kresoli ≥98.0% (by GC)
3-Nitro-o-kresoli
Kaava: C₇H₇NO₃
Sulamispiste: 149…152 °C
MDL numero: MFCD00007241
CAS-numero: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffd4881cc438070ca80cb63544811fa0e79c1069&vid=ffd4881cc438070ca80cb63544811fa0e79c1069&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR