Norbornadiene rhodium(I) chloride dimer

Toimittaja: TCI

N0453-100MG
TCIAN0453-100MGEA 123 EUR
TCIAN0453-100MG
Norbornadiene rhodium(I) chloride dimer
Norbornadiene rhodium(I) chloride dimer
Kaava: C₁₄H₁₆Cl₂Rh₂
MDL numero: MFCD00198060
CAS-numero: 12257-42-0
EINECS: 235-510-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b73f2fc7d16685070dbd64699db2cd62a5fa0fc&vid=d9053ff3c52394124b6d4eee7a12f04c3b4d30c7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Elemental analysis (Carbon) 35.70 to 37.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR