4-Nitrosophenol ≥98.0% (by titrimetric analysis) wetted with ca. 40% water (unit weight on dry weight basis)

Toimittaja: TCI
Vaara

N0374-25G
TCIAN0374-25GEA 96.8 EUR
TCIAN0374-25G
4-Nitrosophenol ≥98.0% (by titrimetric analysis) wetted with ca. 40% water (unit weight on dry weight basis)
4-Nitrosophenol
Kaava: C₆H₅NO₂
MDL numero: MFCD00002329
CAS-numero: 104-91-6
EINECS: 203-251-6
UN: 1325
ADR: 4.1,II
Merck Index: 14,06642

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR