Neostigmine bromide ≥99.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

N0358-25G N0358-5G
TCIAN0358-25GEA 175 EUR
TCIAN0358-25G TCIAN0358-5G
Neostigmine bromide ≥99.0% (by titrimetric analysis)
Neostigmine bromide
Kaava: C₁₂H₁₉BrN₂O₂
MDL numero: MFCD00011795
CAS-numero: 114-80-7
EINECS: 204-054-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,06464

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 % (after drying)
Solubility in Water almost transparency
Drying loss max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR