Methyl pelargonate ≥96.0% (by GC)

Toimittaja: TCI

Synonyms: Methyl nonanoate, Nonanoic acid methyl ester, Pelargonic acid methyl ester

N0290-25ML
TCIAN0290-25MLEA 72.6 EUR
TCIAN0290-25ML
Methyl pelargonate ≥96.0% (by GC)
Methyl pelargonate
Kaava: C₁₀H₂₀O₂
Kiehumispiste: 214°C
Leimahduspiste: 87°C
MDL numero: MFCD00009569
CAS-numero: 1731-84-6
EINECS: 217-052-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59604bc8f5bc395784700bfbe6d39873f4144818&vid=59604bc8f5bc395784700bfbe6d39873f4144818&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR