trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M3219-1G M3219-5G
TCIAM3219-1GEA 75.9 EUR
TCIAM3219-1G TCIAM3219-5G
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by GC)
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid
Kaava: C₁₂H₁₅NO₄
Storage Temperature: Pakastin
MDL numero: MFCD11519178
CAS-numero: 69907-67-1

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR