(±)-Methyl-2,3-dibromopropionate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M3182-25G M3182-5G
TCIAM3182-25GEA 82.5 EUR
TCIAM3182-25G TCIAM3182-5G
(±)-Methyl-2,3-dibromopropionate ≥98.0% (by GC)
(±)-Methyl-2,3-dibromopropionate
Kaava: C₄H₆Br₂O₂
Kiehumispiste: 204°C
MDL numero: MFCD00017882
CAS-numero: 1729-67-5
EINECS: 217-044-3

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR