2-Metyyli-1-tetraloni ≥98.0% (by GC)

Toimittaja: TCI

M3159-1G M3159-5G
TCIAM3159-1GEA 41.8 EUR
TCIAM3159-1G TCIAM3159-5G
2-Metyyli-1-tetraloni ≥98.0% (by GC)
2-Metyyli-1-tetraloni
Kaava: C₁₁H₁₂O
Kiehumispiste: 131°C
Sulamispiste: 15°C
MDL numero: MFCD00001690
CAS-numero: 1590-08-5
EINECS: 216-461-8

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR