4-Methyl-2-(p-tolyl)pyridine ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

M3105-200MG M3105-1G
TCIAM3105-200MGEA 33 EUR
TCIAM3105-200MG TCIAM3105-1G
4-Methyl-2-(p-tolyl)pyridine ≥98.0% (by GC, titration analysis)
4-Methyl-2-(p-tolyl)pyridine
Kaava: C₁₃H₁₃N
MDL numero: MFCD09031945
CAS-numero: 80635-92-3

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR