1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

M3059-5G M3059-25G
TCIAM3059-5GEA 37.4 EUR
TCIAM3059-5G TCIAM3059-25G
1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide
Kaava: C₉H₁₃F₆N₃O₄S₂
MDL numero: MFCD15071469
CAS-numero: 216299-72-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b2ce849069240868e3e97102b599e25008926e&vid=66ae2c81480a51d1ed6fdc477d1dd0fb814e642c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR