4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98.0% (by HPLC)

Toimittaja: TCI

M3028-100MG M3028-500MG
TCIAM3028-100MGEA 169 EUR
TCIAM3028-100MG TCIAM3028-500MG
4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl-Β-D-cellobiopyranoside
Kaava: C₂₂H₂₈O₁₃
Storage Temperature: Jääkaappi
MDL numero: MFCD00063279
CAS-numero: 72626-61-0
EINECS: 276-744-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dde3ec816047dfdf231976861a8513642a4aada9&vid=dde3ec816047dfdf231976861a8513642a4aada9&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -92.0 to -97.0 deg (C=0.2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR