Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M2863-1G M2863-5G
TCIAM2863-1GEA 106 EUR
TCIAM2863-1G TCIAM2863-5G
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate ≥98.0% (by GC)
Methyl-1,5-dimethyl-1H-pyrazole-3-carboxylate
Kaava: C₇H₁₀N₂O₂
Kiehumispiste: 144°C
MDL numero: MFCD06661312
CAS-numero: 10250-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b4fdc3778369a33441ba0d1f7a26148d3ef197&vid=34b4fdc3778369a33441ba0d1f7a26148d3ef197&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 73.0 to 77.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR