Methyl-6-hydroxynicotinate ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

M2727-1G M2727-5G
TCIAM2727-1GEA 58.3 EUR
TCIAM2727-1G TCIAM2727-5G
Methyl-6-hydroxynicotinate ≥97.0% (by GC)
Methyl-6-hydroxynicotinate
Kaava: C₇H₇NO₃
MDL numero: MFCD04113616
CAS-numero: 66171-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e266128323fe5fa44e1f7af440feb65bfb419ea&vid=4e266128323fe5fa44e1f7af440feb65bfb419ea&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR