2-Methyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

M2626-1G M2626-5G
TCIAM2626-1GEA 84.7 EUR
TCIAM2626-1G TCIAM2626-5G
2-Methyl-5-thiazolecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Methyl-5-thiazolecarboxylic acid
Kaava: C₅H₅NO₂S
MDL numero: MFCD01760300
CAS-numero: 40004-69-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be7aecc0549982a5246d319bd4f4c586eb356206&vid=be7aecc0549982a5246d319bd4f4c586eb356206&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR