3-Metyyli-1-buteeni ∼15% ∼2.5 mol/L in dichloromethane

Toimittaja: TCI
Vaara

M2563-100ML M2563-500ML
TCIAM2563-100MLEA 62.7 EUR
TCIAM2563-100ML TCIAM2563-500ML
3-Metyyli-1-buteeni ∼15% ∼2.5 mol/L in dichloromethane
3-Metyyli-1-buteeni
Kaava: C₅H₁₀
Storage Temperature: Jääkaappi
MDL numero: MFCD00008937
CAS-numero: 563-45-1
EINECS: 209-249-1
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70de3d887801771db74b3578806902eeaac8551&vid=f70de3d887801771db74b3578806902eeaac8551&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 15.0% to 17.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR