Methyl-5-bromo-2-methylbenzoate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M2542-1G M2542-5G
TCIAM2542-1GEA 18.7 EUR
TCIAM2542-1G TCIAM2542-5G
Methyl-5-bromo-2-methylbenzoate ≥98.0% (by GC)
Methyl-5-bromo-2-methylbenzoate
Kaava: C₉H₉BrO₂
Kiehumispiste: 265°C
MDL numero: MFCD09839555
CAS-numero: 79669-50-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb44f6d1141a011c024880fbcaf2ec03e32b1e87&vid=bb44f6d1141a011c024880fbcaf2ec03e32b1e87&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR