Methyl-2-bromoisonicotinate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M2461-1G M2461-5G
TCIAM2461-1GEA 50.6 EUR
TCIAM2461-1G TCIAM2461-5G
Methyl-2-bromoisonicotinate ≥98.0% (by GC)
Methyl-2-bromoisonicotinate
Kaava: C₇H₆BrNO₂
MDL numero: MFCD03791265
CAS-numero: 26156-48-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bc45f3cb7e2d03482f486f93df43b5e9a57c502&vid=9bc45f3cb7e2d03482f486f93df43b5e9a57c502&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 34.0 to 38.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR