4-Methyl-2-hexylamine hydrochloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

M2409-1G M2409-5G M2409-25G
TCIAM2409-1GEA 18.7 EUR
TCIAM2409-1G TCIAM2409-5G TCIAM2409-25G
4-Methyl-2-hexylamine hydrochloride ≥98.0% (by titrimetric analysis)
4-Methyl-2-hexylamine hydrochloride
Kaava: C₇H₁₇N
MDL numero: MFCD09038354
CAS-numero: 13803-74-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e75852d57b3a94841a88ebd36fb8f1c43ed9fb1&vid=9e75852d57b3a94841a88ebd36fb8f1c43ed9fb1&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR