Methyl-2-bromohexanoate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M2378-25G M2378-250G
TCIAM2378-25GEA 17.6 EUR
TCIAM2378-25G TCIAM2378-250G
Methyl-2-bromohexanoate ≥98.0% (by GC)
Methyl-2-bromohexanoate
Kaava: C₇H₁₃BrO₂
Kiehumispiste: 94°C
Leimahduspiste: 69°C
MDL numero: MFCD00045038
CAS-numero: 5445-19-2
EINECS: 226-643-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b62bd2f809350b848e2badee67893e62dd4c7f9b&vid=b62bd2f809350b848e2badee67893e62dd4c7f9b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR