Methyl D-3-phenyllactate ≥98.0% (by GC)

Toimittaja: TCI

M2350-1G M2350-5G
TCIAM2350-1GEA 18.7 EUR
TCIAM2350-1G TCIAM2350-5G
Methyl D-3-phenyllactate ≥98.0% (by GC)
Methyl D-3-phenyllactate
Kaava: C₁₀H₁₂O₃
Kiehumispiste: 115°C
MDL numero: MFCD09842268
CAS-numero: 27000-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fd10037121605bface13bcfc789319df0c34ecf&vid=0fd10037121605bface13bcfc789319df0c34ecf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C
Specific rotation [a]20/D -4.0 to -6.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR