Methyl-2-(chloromethyl)benzoate ≥97.0% (by GC)

Toimittaja: TCI
Vaara

M2203-25G M2203-5G
TCIAM2203-25GEA 497 EUR
TCIAM2203-25G TCIAM2203-5G
Methyl-2-(chloromethyl)benzoate ≥97.0% (by GC)
Methyl-2-(chloromethyl)benzoate
Kaava: C₉H₉ClO₂
MDL numero: MFCD06200781
CAS-numero: 34040-62-5
EINECS: 251-805-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=363f5303732f3703f91b5fc7286e40582148e93b&vid=363f5303732f3703f91b5fc7286e40582148e93b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR