N-Methyl-3-pyrrolidinol ≥97.0% (by titrimetric analysis)

Toimittaja: TCI

M2115-5G M2115-25G
TCIAM2115-5GEA 81.4 EUR
TCIAM2115-5G TCIAM2115-25G
N-Methyl-3-pyrrolidinol ≥97.0% (by titrimetric analysis)
N-Methyl-3-pyrrolidinol
Kaava: C₅H₁₁NO
Kiehumispiste: 75°C
Leimahduspiste: 71°C
MDL numero: MFCD00003176
CAS-numero: 13220-33-2
EINECS: 236-189-3

Order Now

Testitulosten erittely

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR