2-(Metyylitio)bentsaldehydi ≥97.0% (by GC)

Toimittaja: TCI

M2094-1G M2094-5G
TCIAM2094-1GEA 139 EUR
TCIAM2094-1G TCIAM2094-5G
2-(Metyylitio)bentsaldehydi ≥97.0% (by GC)
2-(Metyylitio)bentsaldehydi
Kaava: C₈H₈OS
Kiehumispiste: 149°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00196822
CAS-numero: 7022-45-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c345e1ffb4e5389755968e897af53e641a34eced&vid=c345e1ffb4e5389755968e897af53e641a34eced&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR