N-Methyliminodiacetic acid ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

M2090-5G M2090-25G
TCIAM2090-5GEA 17.6 EUR
TCIAM2090-5G TCIAM2090-25G
N-Methyliminodiacetic acid ≥98.0% (by GC, titration analysis)
N-Methyliminodiacetic acid
Kaava: C₅H₉NO₄
Sulamispiste: 219°C
MDL numero: MFCD00004284
CAS-numero: 4408-64-4
EINECS: 224-557-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc892a2e05448b8fbf39ed3cd1daa33e5fb5a2d&vid=5cc892a2e05448b8fbf39ed3cd1daa33e5fb5a2d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR