Methyl thiosalicylate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M1881-25G M1881-5G
TCIAM1881-25GEA 230 EUR
TCIAM1881-25G TCIAM1881-5G
Methyl thiosalicylate ≥98.0% (by GC)
Methyl thiosalicylate
Kaava: C₈H₈O₂S
Kiehumispiste: 123°C
MDL numero: MFCD00060678
CAS-numero: 4892-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d16458bc0461bda8f11e9d283e65fa200c6c165&vid=0d16458bc0461bda8f11e9d283e65fa200c6c165&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR