Methyl-2-(p-tolyl)benzoate ≥98.0% (by GC)

Toimittaja: TCI

M1809-25G M1809-5G
TCIAM1809-25GEA 146 EUR
TCIAM1809-25G TCIAM1809-5G
Methyl-2-(p-tolyl)benzoate ≥98.0% (by GC)
Methyl-2-(p-tolyl)benzoate
Kaava: C₁₅H₁₄O₂
MDL numero: MFCD03453661
CAS-numero: 114772-34-8

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR