Methyl-2-ethoxybenzoate ≥99.0% (by GC)

Toimittaja: TCI

M1785-25G
TCIAM1785-25GEA 124 EUR
TCIAM1785-25G
Methyl-2-ethoxybenzoate ≥99.0% (by GC)
Methyl-2-ethoxybenzoate
Kaava: C₁₀H₁₂O₃
Kiehumispiste: 245°C
MDL numero: MFCD00017261
CAS-numero: 3686-55-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b047753517d98bdb9036a62e880a1b3a14fe68a0&vid=b047753517d98bdb9036a62e880a1b3a14fe68a0&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR