Metyyli-3-merkaptopropionaatti ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M1434-25G M1434-500G
TCIAM1434-25GEA 46.2 EUR
TCIAM1434-25G TCIAM1434-500G
Metyyli-3-merkaptopropionaatti ≥98.0% (by GC)
Metyyli-3-merkaptopropionaatti
Kaava: C₄H₈O₂S
Kiehumispiste: 54 °C (1013 hPa)
Sulamispiste: –60 °C
Tiheys: 1,085 g/cm³ (20 °C)
Leimahduspiste: 64°C
MDL numero: MFCD00004895
CAS-numero: 2935-90-2
EINECS: 220-912-4
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8aa77197e413a2d8504cf31235bf0fe170dc8968&vid=8aa77197e413a2d8504cf31235bf0fe170dc8968&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR