(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic acid ≥99.0% (by HPLC)

Toimittaja: TCI
Varoitus

M1367-100MG
TCIAM1367-100MGEA 198 EUR
TCIAM1367-100MG
(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic acid ≥99.0% (by HPLC)
(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic acid
Kaava: C₁₄H₁₄O₃
MDL numero: MFCD03093634
CAS-numero: 102691-93-0
Merck Index: 14,06224

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Optical purity (LC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR