4-Metyylibentsofenoni ≥95.0% (by GC)

Toimittaja: TCI

M1248-25G M1248-250G
TCIAM1248-25GEA 18.7 EUR
TCIAM1248-25G TCIAM1248-250G
4-Metyylibentsofenoni ≥95.0% (by GC)
4-Metyylibentsofenoni
Kaava: C₁₄H₁₂O
Kiehumispiste: 328°C
Sulamispiste: 53…57 °C
Tiheys: 0,9926 g/cm³ (20 °C)
MDL numero: MFCD00008553
CAS-numero: 134-84-9
EINECS: 205-159-1
Merck Index: 12,07470
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb005239a4a60d9e8505ba0b44f1e4e9dd119a06&vid=cb005239a4a60d9e8505ba0b44f1e4e9dd119a06&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 54.0 to 58.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR