Methyl-3-methoxypropionate ≥99.0% (by GC)

Toimittaja: TCI
Varoitus

M0962-25ML M0962-500ML
TCIAM0962-25MLEA 17.6 EUR
TCIAM0962-25ML TCIAM0962-500ML
Methyl-3-methoxypropionate ≥99.0% (by GC)
Methyl-3-methoxypropionate
Kaava: C₅H₁₀O₃
Kiehumispiste: 144°C
Leimahduspiste: 47°C
MDL numero: MFCD00059085
CAS-numero: 3852-09-3
EINECS: 223-358-1
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4921ee19f02ed988eba9acce3c8012709fe097b6&vid=4921ee19f02ed988eba9acce3c8012709fe097b6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR