4-Methyl-4-heptanol ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

M0954-5ML
TCIAM0954-5MLEA 215 EUR
TCIAM0954-5ML
4-Methyl-4-heptanol ≥98.0% (by GC)
4-Methyl-4-heptanol
Kaava: C₈H₁₈O
Kiehumispiste: 62°C
Leimahduspiste: 52°C
MDL numero: MFCD00021840
CAS-numero: 598-01-6
UN: 1987
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81120aaa28c44fca6e67b818a67d60826ea5f496&vid=81120aaa28c44fca6e67b818a67d60826ea5f496&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR