5-Methylthiophene-2-carboxaldehyde ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

M0911-5G M0911-25G
TCIAM0911-5GEA 18.7 EUR
TCIAM0911-5G TCIAM0911-25G
5-Methylthiophene-2-carboxaldehyde ≥97.0% (by GC)
5-Methylthiophene-2-carboxaldehyde
Kaava: C₆H₆OS
Kiehumispiste: 205°C
Leimahduspiste: 87°C
MDL numero: MFCD00005434
CAS-numero: 13679-70-4
EINECS: 237-178-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84d978d50c19680844d452ee5b65102ad3a6ce4c&vid=84d978d50c19680844d452ee5b65102ad3a6ce4c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Yellow to Amber to Dark purple clear liquid to cloudy liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR